Order Entry
Austria
ContactUsLinkComponent
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine for synthesis
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine for synthesis
Catalog # 8.14399.0001
Supplier:  Merck
CAS Number:  
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine for synthesis
Catalog # 8.14399.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 ml
  • Seq:
    2070

Specifications

About this item

 :
 :257.16 g/mol
CAS Number:31886-58-5
Formula:C₁₄H₁₉FeN
MDL Number:MFCD00066273