Acetonitrile with 0.1% (v/v) trifluoroacetic acid ≥99.5%, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Catalog # 34976-2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Assay (GC):Min. 99.5 %
- Assay trifluoroacetic acid:0.093 - 0.107 Vol.%
- Calcium (Ca):Max. 0.5 ppm
- Potassium (K):Max. 0.5 ppm
- Magnesium (Mg):Max. 0.5 ppm
- Sodium (Na):Max. 2 ppm
- Transmittance at 210 nm:Min. 30 %
- Transmittance at 230 nm:Min. 50 %
- Transmittance from 254 nm:Min. 90 %
- HPLC Gradient at 254 nm:Max. 50 mAU
- LC-MS application:complying
Specification Test Results
About this item
:
Boiling Pt:81…82 °C
Density:0.78 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
:GHS02
:GHS08
:GHS07
CAS Number:76-05-1
:75-05-8
MDL Number:MFCD00001878