Order Entry
Austria
ContactUsLinkComponent
4,4'-Methylenebis(3-chloro-2,6-diethylaniline) for synthesis, Sigma-Aldrich®
4,4'-Methylenebis(3-chloro-2,6-diethylaniline) for synthesis, Sigma-Aldrich®
Catalog # 8.14676.0025
Supplier:  Merck
CAS Number:  
4,4'-Methylenebis(3-chloro-2,6-diethylaniline) for synthesis, Sigma-Aldrich®
Catalog # 8.14676.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    88 - 90 °C
  • Molar Mass:
    379.37 g/mol
  • Bulk Density:
    610 - 650 kg/m3

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
 :379.3664 g/mol
EINECS:402-130-7
REACH:01-0000015192-78
CAS Number:106246-33-7
Formula:C₂₁H₂₈Cl₂N₂
MDL Number:MFCD00071551