Order Entry
Austria
ContactUsLinkComponent
Yeast invertase
Yeast invertase
Catalog # I9274-100MG
Supplier:  Merck
CAS Number:  
undefined
Yeast invertase
Catalog # I9274-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • CAS No.:
    9001-57-4
  • Species:
    Yeast
  • Enzyme name:
    Invertase

Specifications

About this item