Order Entry
Austria
ContactUsLinkComponent
 
2-Hydroxy-2-methylpropiophenone
Catalog # EHERC14233500
Supplier:  EHRENSTORFER
CAS Number:  
2-Hydroxy-2-methylpropiophenone
Catalog # EHERC14233500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :164.2000 g/mol
EINECS:231-272-0
CAS Number:7473-98-5
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00040710