(4-Aminophenyl)acetonitrile, Sigma-Aldrich®
Catalog # SIALA42050-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:132.17 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:222-587-4
UN:3439
CAS Number:3544-25-0
Formula:C₈H₈N₂
MDL Number:MFCD00007912