Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
Tetrahydrofuran ≥99.9% unstabilised, SPECTRONORM® for spectroscopy
 
undefined
 
 
 
Tetrahydrofuran ≥99.9% unstabilised, SPECTRONORM® for spectroscopy
 

 

  • Assay (GC):
    ≥ 99,9 %
  • Acidity:
    ≤ 0,0002 meq/g
  • Alkalinity:
    ≤ 0,0002 meq/g
  • Colouration:
    ≤ 10 APHA
  • Evaporation residue:
    ≤ 2 ppm
  • Water:
    ≤ 100 ppm
  • Absorbance (215 nm):
    ≤ 0,52
  • Absorbance (245 nm):
    ≤ 0,30
  • Absorbance (265 nm):
    ≤ 0,10
  • Absorbance (275 nm):
    ≤ 0,05
  • Absorbance (from 310 nm):
    ≤ 0,01
  • Transmittance (215 nm):
    ≥ 30 %
  • Transmittance (245 nm):
    ≥ 50 %
  • Transmittance (265 nm):
    ≥ 80 %
  • Transmittance (275 nm):
    ≥ 90 %
  • Transmittance (from 310 nm):
    ≥ 98 %
  • Fluorescence (as quinine) (254 nm):
    ≤ 1,0 ppb
  • Fluorescence (as quinine) (365 nm):
    ≤ 1,0 ppb

 

Frequently Bought Together