Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide, BAKER ANALYZED® ACS, J.T.Baker®
Corrosive
Sodium hydroxide, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
Corrosive
Sodium hydroxide, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 97.0 %
  • Calcium (Ca):
    max. 0.005 %
  • Chloride (Cl):
    max. 0.005 %
  • Potassium (K):
    max. 0.01 %
  • Magnesium (Mg):
    max. 0.002 %
  • Heavy Metals (as Ag):
    max. 0.001 %
  • Sodium Carbonate (Na2CO3):
    max. 1.0 %
  • Iron (Fe):
    max. 3
  • Mercury (Hg):
    max. 0.1
  • Nickel (Ni):
    max. 5
  • Nitrogen Compounds (as N):
    max. 3
  • Phosphate (PO4):
    max. 2
  • Sulfate (SO4):
    max. 5

Specification Test Results

Related Information

Frequently Bought Together