Order Entry
Austria
ContactUsLinkComponent
DL-α-Tocopherol acetate, Supelco®
DL-α-Tocopherol acetate, Supelco®
Catalog # PHR1030-500MG
Supplier:  Merck
CAS Number:  
undefined
DL-α-Tocopherol acetate, Supelco®
Catalog # PHR1030-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :472.75 g/mol
EINECS:231-710-0
CAS Number:7695-91-2
 :52225-20-4
Formula:C₃₁H₅₂O₃
MDL Number:MFCD00072042