



n-Heptane, BAKER, Laboratory Reagent, J.T.Baker®
Artikelnr. 8270.9200ST
Supplier: Avantor
CAS Number:
Specifications
- Pk:200 L
- Seq:5549
Specifications
Specification Test Results
- Assay (by GC):min. 95 %
- Boiling Point:98 - 99 °C
- Residue after Evaporation:max. 0.001 %
Specification Test Results
About this item
:
:100.20 g/mol
Merck Index:13,04679
Boiling Pt:98.4 °C (1013 hPa)
Melting Pt:–90.6 °C
Density:0.6838 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544