Order Entry
Austria
ContactUsLinkComponent
2-Propanol ≥99.8%, SPECTRONORM® for spectroscopy
Flammable
Irritant
2-Propanol ≥99.8%, SPECTRONORM® for spectroscopy
Catalog # 84706.320
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Irritant
2-Propanol ≥99.8%, SPECTRONORM® for spectroscopy
Catalog # 84706.320
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Assay (GC):
    ≥ 99.9 %
  • Solubility in water:
    Passes test ACS
  • Acidity:
    ≤ 0.0001 meq/g
  • Alkalinity:
    ≤ 0.0001 meq/g
  • Colouration:
    ≤ 10 APHA
  • Carbonyl compounds (as acetone):
    ≤ 20 ppm
  • Carbonyl compounds (as propionaldehyde):
    ≤ 20 ppm
  • Evaporation residue:
    ≤ 5 ppm
  • Water:
    ≤ 0.05 %
  • Absorbance (210 nm):
    ≤ 0.52
  • Absorbance (220 nm):
    ≤ 0.19
  • Absorbance (230 nm):
    ≤ 0.10
  • Absorbance (245 nm):
    ≤ 0.05
  • Absorbance (250 nm):
    ≤ 0.02
  • Absorbance (from 260 nm):
    ≤ 0.01
  • Transmittance (210 nm):
    ≥ 30 %
  • Transmittance (220 nm):
    ≥ 65 %
  • Transmittance (230 nm):
    ≥ 80 %
  • Transmittance (245 nm):
    ≥ 90 %
  • Transmittance (250 nm):
    ≥ 95 %
  • Transmittance (from 260 nm):
    ≥ 98 %
  • Fluorescence (as quinine) (254 nm):
    ≤ 1.0 ppb
  • Fluorescence (as quinine) (365 nm):
    ≤ 1.0 ppb

Specification Test Results

About this item

 :
 :60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
REACH:01-2119457558-25
 :GHS02
 :GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674