Order Entry
Austria
ContactUsLinkComponent
 
 
 
Dichloromethane, BAKER ANALYZED® HPLC for HPLC, J.T.Baker®
 :  Avantor
 
undefined
 
 
Dichloromethane, BAKER ANALYZED® HPLC for HPLC, J.T.Baker®
 :  Avantor
 

 

  • Assay (by GC):
    min. 99.8 %
  • Density (g/ml) at 25°C:
    1.315 - 1.321
  • Chloride (Cl):
    max. 10 ppm
  • Preservative (Amylene) (ppm):
    about 20
  • Residue after Evaporation (in ppm):
    max. 2
  • Titrable Acid (meq/g):
    max. 0.0003
  • Water (H2O):
    max. 0.02 %
  • at 254 nm:
    max. 0.01
  • at 280 nm:
    max. 0.01
  • at 350 nm:
    max. 0.01
  • UV Cut-off, nm:
    max. 233

 

Frequently Bought Together