Pronase E
Marketsource
Supplier: ROCHE
About this item
2 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ROCH10165921001
- ROCH11459643001
- Enzyme name:
- Pronase E
- Pronase E
- MDL No.:
- MFCD02253017
- MFCD02253017
- CAS No.:
- 9036-06-0
- 9036-06-0
- Storage conditions:
- Freezer
- Freezer
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:Marketsource
More
Specifications
Cat. No.ROCH10165921001Enzyme namePronase EMDL No.MFCD02253017CAS No.9036-06-0Storage conditionsFreezerMarketsource - Specifications:Marketsource
More
Specifications
Cat. No.ROCH11459643001Enzyme namePronase EMDL No.MFCD02253017CAS No.9036-06-0Storage conditionsFreezerMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



