Order Entry
Austria
ContactUsLinkComponent
Acetonitrile ≥99.9%, SPECTRONORM® for spectroscopy
Acetonitrile ≥99.9%, SPECTRONORM® for spectroscopy
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Acetonitrile ≥99.9%, SPECTRONORM® for spectroscopy
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay (GC):
    ≥ 99,9 %
  • Acidity:
    ≤ 0,0002 meq/g
  • Alkalinity:
    ≤ 0,0001 meq/g
  • Colouration:
    ≤ 10 APHA
  • Evaporation residue:
    ≤ 2 ppm
  • Water:
    ≤ 100 ppm
  • Absorbance (190 nm):
    ≤ 0,70
  • Absorbance (195 nm):
    ≤ 0,22
  • Absorbance (200 nm):
    ≤ 0,05
  • Absorbance (215 nm):
    ≤ 0,02
  • Absorbance (from 230 nm):
    ≤ 0,01
  • Transmittance (190 nm):
    ≥ 20 %
  • Transmittance (195 nm):
    ≥ 60 %
  • Transmittance (200 nm):
    ≥ 90 %
  • Transmittance (215 nm):
    ≥ 95 %
  • Transmittance (from 230 nm):
    ≥ 98 %
  • Fluorescence (as quinine) (254 nm):
    ≤ 0,5 ppb
  • Fluorescence (as quinine) (365 nm):
    ≤ 0,5 ppb

Specification Test Results

Frequently Bought Together