Order Entry
Austria
ContactUsLinkComponent
Trisodium phosphate dodecahydrate 98.0-102.0%, crystals, AR® ACS, Macron Fine Chemicals™
Irritant
Trisodium phosphate dodecahydrate 98.0-102.0%, crystals, AR® ACS, Macron Fine Chemicals™
Supplier:  Avantor
Certificates
undefined
Irritant
Trisodium phosphate dodecahydrate 98.0-102.0%, crystals, AR® ACS, Macron Fine Chemicals™
Supplier:  Avantor

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
  • ACS - Assay (Na₃PO₄·12H₂O):
    98.0 - 102.0 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Excess Alkali (as NaOH):
    ≤2.5 %
  • ACS - Heavy Metals (as Pb):
    ≤0.001 %
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Iron (Fe):
    ≤0.001 %
  • ACS - Sulfate (SO₄):
    ≤0.01 %
  • Appearance (colorless or white crystals):

Specification Test Results

Frequently Bought Together