Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid 32%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Corrosive
Irritant
Hydrochloric acid 32%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Corrosive
Irritant
Hydrochloric acid 32%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 32 %
  • Ammonium (NH4):
    max. 1 ppm
  • Free Chlorine:
    max. 0.5 ppm
  • Residue after Ignition:
    max. 5 ppm
  • Sulfate (SO4):
    max. 1 ppm
  • Sulfite (SO3):
    max. 2 ppm
  • Silver (Ag):
    max. 0.1
  • Aluminium (Al):
    max. 0.5
  • Arsenic (As):
    max. 0.1
  • Barium (Ba):
    max. 0.1
  • Beryllium (Be):
    max. 0.1
  • Calcium (Ca):
    max. 0.5
  • Cadmium (Cd):
    max. 0.1
  • Cobalt (Co):
    max. 0.1
  • Chromium (Cr):
    max. 0.1
  • Copper (Cu):
    max. 0.1
  • Iron (Fe):
    max. 0.5
  • Germanium (Ge):
    max. 0.1
  • Potassium (K):
    max. 0.1
  • Lithium (Li):
    max. 0.1
  • Magnesium (Mg):
    max. 0.5
  • Manganese (Mn):
    max. 0.1
  • Molybdenum (Mo):
    max. 0.1
  • Sodium (Na):
    max. 0.5
  • Nickel (Ni):
    max. 0.1
  • Lead (Pb):
    max. 0.1
  • Strontium (Sr):
    max. 0.1
  • Titanium (Ti):
    max. 0.1
  • Thallium (Tl):
    max. 0.1
  • Vanadium (V):
    max. 0.1
  • Zinc (Zn):
    max. 0.5
  • Zirconium (Zr):
    max. 0.1

Specification Test Results

Related Information

Frequently Bought Together