4,4'-(1-Methylpropylidene)diphenol
Catalog # EHERC10655670
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:242.32 g/mol
Merck Index:12,01339
Boiling Pt:412 °C (1013 hPa)
Melting Pt:124…125 °C
EINECS:201-025-1
CAS Number:77-40-7
Formula:C₁₆H₁₈O₂
MDL Number:MFCD00020179