ContactUsLinkComponent
Western blotting detection reagent, Amersham™ ECL Select™
Western blotting detection reagent, Amersham™ ECL Select™
Catalog # RPN2235
Supplier:  Cytiva
CAS Number:  
undefined
Western blotting detection reagent, Amersham™ ECL Select™
Catalog # RPN2235
Supplier:  Cytiva
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Amersham™ ECL Select™ Western blotting detection reagent for 1000 cm² membrane
Specifications
About this item
Amersham ECL Select is a high sensitivity reagent for chemiluminescent Western blotting detection.
  • Exceptional signal intensity resulting in brighter, clearer bands at low protein levels
  • For detection of low to very low abundant proteins
  • Quantitative measurements
  • Works with a wide range of antibody dilutions
  • Optimized for use with Amersham Hybond P (PVDF) membranes and compatible with Amersham Protran (nitrocellulose) membranes
  • Optimized for CCD-based imaging, for example Amersham Imager 600 or ImageQuant LAS 500, and compatible with X-ray film such as Amersham Hyperfilm ECL
  • Compatible with Amersham ECL Molecular Weight Rainbow Markers and Amersham ECL DualVue Western Blotting Markers
  • Optimized for use with Amersham Hybond-P (PVDF) membranes and compatible with Amersham Hybond ECL (nitrocellulose) membranes
Signals can be detected with CCD camera or X-ray film. Amersham ECL Select typically exhibits a dynamic range that is complementary to Amersham.
High signal intensity makes Amersham ECL Select suitable for demanding Western blotting applications, allowing detection of minute protein quantities. Its long lasting signals allows for multiple exposures and a convenient time window between the experiment and analysis. Signals can be detected with a CCD camera or X-ray film. Delivering high sensitivity even when using highly diluted primary and secondary antibodies.