ContactUsLinkComponent
Beakers, low or tall form
Beakers, low or tall form
Catalog # KAVA632417012940
Supplier:  Kavalier
Supplier #: 632417012940
undefined
Beakers, low or tall form
Catalog # KAVA632417012940
Supplier:  Kavalier
Supplier #: 632417012940
Log in for availability
Select an Option
95 mm
Select an Option
1000 ml
Select an Option
180 mm
Select an Option
39
Select an Option
10
About this item
Borosilicate glass, grade 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Autoclavable
Ideal for general laboratory use. With spout and white graduations.
Certifications: DIN 12331 (exept 25 ml/5000 ml tall form and 10000 ml low form supplementary to DIN). ISO 3819.
Frequently Bought Together