ContactUsLinkComponent
DNA-Dye NonTox Nucleic acid gel stain
DNA-Dye NonTox Nucleic acid gel stain
  A9555.1000
 :  
undefined
DNA-Dye NonTox Nucleic acid gel stain
  A9555.1000
 :  
  • Pk:
    1 mL
  • Description:
    DNA-Dye NonTox
DNA-Dye NonTox is a highly sensitive, non toxic, fluorescent staining reagent for the detection of double-stranded DNA (dsDNA). The dye produces instant visualisation of DNA bands on gels upon blue light or UV illumination.
  • nonhazardous Ethidium bromide substitute
  • Loading buffer with highly sensitive DNA dye
  • Higher transfection rates since DNA structure and integrity is not affected
  • Ready to use
  • Contains three tracking dyes Bromophenol Blue, Xylene Cyanol FF, and Orange G for visually tracking the DNA migration during the electrophoresis process
DNA-Dye NonTox is ideal in terms of environmental safety by offering a non hazardous alternative to ethidium bromide. In addition, the dye included in DNA-Dye NonTox does not affect the structure and integrity of DNA.
 : Supplied in 6X DNA loading buffer to prepare DNA markers and samples for loading on agarose or polyacrylamide gels.