PIG® Extra-Duty universal mat pads and rolls
Lieferant: NEW PIG
Zertifikate
Über diesen Artikel
Both sides of this absorbent mat are tough enough for rough surfaces. PIG Extra-Duty Mat is highly abrasion resistant so it won't rip, tear or fray.
- Special spunbond outer layers resist ripping and fraying from wiping rough surfaces or parts that would shred conventional mats
- Eight layers of 100% polypropylene are thermally bonded; won't rip, tear or fray even when saturated
- Highly absorbent, fine-fibre construction won't leave behind liquids or fibre residue
- Absorbs and retains oils, coolants, solvents and water
- Low-lint for wiping rough parts or equipment and for cleaning in paint booth applications; also great for lining benches for rough or sharp parts
- Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat whilst on the job
Rolls are perforated every 25 cm; easy to tear to size, use only what you need. Heavy-weight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups. 25% or more recycled content.
Frequently Bought Together