Orders LinkContactUsLinkComponent
PIG® spill bag
PIG® spill bag
Supplier:   NEW PIG
PIG® spill bag
Supplier:  NEW PIG
Certificates
PIG® spill bag
3 Options Available Below
Product Details & Documents
High-visibility, compact spill bag travels anywhere you do for spill response on the go.
  • Brightly coloured yellow bag features a reflective stripe for greater visibility and easy identification
  • Drawstring closure is ideal for carrying or hanging on a wall
  • Packed with a variety of PIG® absorbents to handle small spills
PIG® spill bag is flexible and small enough to fit behind the seat of a lorry; stows almost anywhere and is easy to carry in the event of a spill
Caution: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.
Product Family Options
Universal
Product Information
  • Absorbency
    W×D×H
    Availability
    Price
  • 121-0066
    Spill bag, includes 20 mat pads, 1 absorbent sock and 2 disposal bags
    Spill bag, includes 20 mat pads, 1 absorbent sock and 2 disposal bags
    10,4 L
    140×250×360 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,80 €
    28,80 €
    1 * 1 ST
       28,80 €
HAZ-MAT
Product Information
  • Absorbency
    W×D×H
    Availability
    Price
  • 121-0070
    Spill bag, includes 20 mat pads, 1 absorbent sock and 2 disposal bags
    Spill bag, includes 20 mat pads, 1 absorbent sock and 2 disposal bags
    10,4 L
    140×250×360 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,00 €
    37,00 €
    1 * 1 ST
       37,00 €
Oil-Only
Product Information
  • Absorbency
    W×D×H
    Availability
    Price
  • 121-0074
    Spill bag, includes 20 mat pads, 1 absorbent sock and 2 disposal bags
    Spill bag, includes 20 mat pads, 1 absorbent sock and 2 disposal bags
    11,3 L
    140×250×360 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,80 €
    34,80 €
    1 * 1 ST
       34,80 €