Orders LinkContactUsLinkComponent
Washers, BOLA
Washers, BOLA
Supplier:   Bohlender
Washers, BOLA
Supplier:  Bohlender
Certificates
Washers, BOLA
2 Options Available Below
Product Details & Documents
PA
  • Temperature resistance: from –40 to +100 °C
For stabilising the flange and avoiding damages when tightening the tube end fitting.
Product Family Options
Product Information
  • Material
    Availability
    Price
  • BOHLF728-16
    for tubing 1,6 mm I-Ø/3,2 mm O-Ø
    for tubing 1,6 mm I-Ø/3,2 mm O-Ø
    PA
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 3,90 €
    3,90 €
    1 * 10 ST
       3,90 €
  • BOHLF728-08
    for tubing 0,8 mm I-Ø/1,6 mm O-Ø
    for tubing 0,8 mm I-Ø/1,6 mm O-Ø
    PA
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 4,00 €
    4,00 €
    1 * 10 ST
       4,00 €