Order Entry
Austria
ContactUsLinkComponent
Tubing holders, BOLA
Tubing holders, BOLA
Catalog # BOHLF706-06
Supplier:  Bohlender
undefined
Tubing holders, BOLA
Catalog # BOHLF706-06
Supplier:  Bohlender

Specifications

  • Description:
    for tubing O. D. 1,6 and 3,2 mm
  • Pk:
    1

Specifications

About this item

For tubing with O.D. 1,6 mm (¹/₁₆") and 3,2 mm (¹/₈").

  • Safe fixing of the tubing during the flanging procedure
  • Easy assembly and handling
  • Injuries due to the hot flanging tips are avoided