ContactUsLinkComponent
Jars with knobbed lid
Jars with knobbed lid
Catalog # HECH44420024
undefined
Jars with knobbed lid
Catalog # HECH44420024
Specifications
  • Ext.Ø:
    100 mm
  • Height:
    100 mm
  • Pk:
    1
Specifications
About this item
Jars made of stainless steel with knob cover.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Easy to clean