Orders LinkContactUsLinkComponent
Separating funnels, conical
Separating funnels, conical
50 ml, 19/26 NS
Catalog # HECH42950024
Separating funnels, conical
Catalog # HECH42950024
Separating funnels, conical
Separating funnels, conical
50 ml, 19/26 NS
Price
42,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Separating funnels, conical, made of DURAN® borosilicate glass 3.3, with solid glass or interchangeable PTFE key and plastic stopper.
  • Large labeling field
  • Excllent temperature resistance
  • Excllent chemical resistance
Certifications: DIN ISO4800
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    50 ml