ContactUsLinkComponent
Separating funnels, pear shaped
Separating funnels, pear shaped
Catalog # HECH42970066
undefined
Separating funnels, pear shaped
Catalog # HECH42970066
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    1000 ml
  • Pk:
    1
Specifications
About this item
Separating funnels, made of borosilicate glass, pear shaped, stopcock with solid glass plug.
  • Without graduations
  • Excellent chemical resistance
  • Excellent temperature resistance