ContactUsLinkComponent
Evaporating flasks
Evaporating flasks
Catalog # IKAA1905700
Supplier:  IKA
Evaporating flasks
Catalog # IKAA1905700
Supplier:  IKA
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    100 ml
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
NS 29/32 standard ground joint socket, pear-shaped, centre neck, thick-walled. Suitable for all standard IKA® rotary evaporators; with white labelling area.