Centrifuge tubes, conical
Catalog # 525-1305
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Closure type:Without cap
- Ø×L:17×98/100 mm
- Type:AR-GLAS®, rimless
- Capacity:15 ml
- Seq:03
- Pk:100
Specifications
About this item
AR®-glass or DURAN® borosilicate glass 3.3, conical.
- Ungraduated
- With rim or rimless