Order Entry
Austria
ContactUsLinkComponent
Dual infrared and core thermometer HACCP, testo 104-IR
Dual infrared and core thermometer HACCP, testo 104-IR
Catalog # 620-1904
Supplier:  Testo
undefined
Dual infrared and core thermometer HACCP, testo 104-IR
Catalog # 620-1904
Supplier:  Testo

Specifications

  • Range:
    −30…+250 °C*
    −50…+250 °C**
  • Resolution:
    0,1 °C
  • Weight:
    165 g
  • W×D×H:
    265×48×19 mm
  • Accuracy:
    ±1,0 (−50…−30,1);
    ±0,5 (−30…+99,9);
    ±1,0% of mV (+100…+250) °C
  • Pk:
    1

Specifications

About this item

This waterproof folding thermometer is a handy aid when carrying out precise food monitoring measurements at any time. It is suitable for applications in production and storage. After use, the probe can be folded away, and the instrument stored safely until the next measurement.

  • Automatic final value recognition thanks to auto hold
  • Storage of minimum/maximum values
  • Rubber coated surface guarantees non slip handling

* Infrared
** NTC

Weight: 165 g

Compliant with Hazard Analysis and Critical Control Points (HACCP) and EN 13485.

Certifications: Compliant with Hazard Analysis and Critical Control Points (HACCP) and EN 13485.
Delivery: Supplied with two AAA batteries for 100 hours of use.