Containers with screw cap, Sisterile®
Supplier: KIMA SAS DI CHIARIN RENZO
Certificates
About this item
Made of PP, translucent.
- Graduated
- Excellent chemical resistance
- Very good temperature resistance
Labelled with writing surface of 28×48 mm and screw cap.
Frequently Bought Together