Order Entry
Austria
ContactUsLinkComponent
 
Capillary columns, Elite-17ht for autosystem
  PERCN9316264
Capillary columns, Elite-17ht for autosystem
  PERCN9316264

 

  • Column Format:
    Capillary
  • Film thickness:
    0,15 µm
  • Ø int.:
    0,25 mm
  • Length:
    30 m
  • Seq:
    03
  • Phase:
    Elite-17ht
  • Pk:
    1

 

 

Elite-17ht columns are extremely versatile, permitting the analysis of pesticides, herbicides, rosin acids, phthalate esters, triglycerides and sterols.