ContactUsLinkComponent
Fluid aspiration system, BioChem VacuuCenter, BVC basic series
Fluid aspiration system, BioChem VacuuCenter, BVC basic series
Supplier:  VACUUBRAND
   Sustainable Options Available
Certificates
undefined
Fluid aspiration system, BioChem VacuuCenter, BVC basic series
Supplier:  VACUUBRAND
   Sustainable Options Available
Specifications
  • Catalog No:
  • 181-0299
  • 181-0298
  • Model:
  • BioChem-VacuuCenter BVC basic G
  • BioChem-VacuuCenter BVC basic
  • Environmentally Preferable:
  • Energy Efficient
  • Energy Efficient
  • Vacuum pump:
  • supplied without pump
  • supplied without pump
  • Final pressure of vacuum pump:
  • 150 mbar
  • 150 mbar
  • Protection class:
  • IP 54
  • IP 54
  • Weight:
  • 2,8 kg
  • 2,8 kg