Order Entry
Austria
ContactUsLinkComponent
Open access transfer station, PuriCare®
Open access transfer station, PuriCare®
Supplier:  LABCONCO
   Sustainable Options Available
Certificates
undefined
Open access transfer station, PuriCare®
Supplier:  LABCONCO
   Sustainable Options Available

Specifications

  • Catalog No:
  • LBCP3820023
  • LBCP3820020
  • LBCP3820033
  • Model:
  • PuriCare®
  • PuriCare®
  • PuriCare®
  • Environmentally Preferable:
  • Sustainable Materials
  • Sustainable Materials
  • Sustainable Materials
  • Filters:
  • 2 HEPA filters 99,99% efficient
  • 2 HEPA filters 99,99% efficient
  • 2 HEPA filters 99,99% efficient
  • Electrical supply:
  • 230 V, 50/60 Hz
  • 230 V, 50 Hz, 7 A
  • 230 V, 50/60 Hz
  • Ext. W×D×H:
  • 1006×762×1946 - 2200 mm
  • 1483×762×1946 - 2200 mm
  • 1232×762×1946 - 2200 mm
  • Int. W×D×H:
  • 1006×760×711 mm
  • 1230×760×710 mm
  • 1232×760×711 mm
  • Weight:
  • 250 kg
  • 272 kg
  • 272 kg

Specifications

Frequently Bought Together