Orders LinkContactUsLinkComponent
Drum funnels
Drum funnels
Supplier:   NEW PIG
Drum funnels
Supplier:  NEW PIG
Certificates
Drum funnels
2 Options Available Below
Product Details & Documents
Thick, double-wall polyethylene construction resists UV rays, corrosion and most chemicals.
  • Allows access to ³/₄" bung for use of liquid-level gauge
  • Level inside surface allows easy draining of containers, parts or filters
  • Zigzag ridges prevent splashes and catch parts or tools
These large diameter funnels covers entire drum top to minimize drips and spills.
Product Family Options
Product Information
  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 135,00 €
    135,00 €
    1 * 1 ST
       135,00 €
  • Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 67,30 €
    67,30 €
    1 * 1 ST
       67,30 €
Accessories
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Pop-up drum fill gauge, prevent overfilling
    Pop-up drum fill gauge, prevent overfilling
    NEWPDRM245
    127 mm
    Log in for availability
    1 * 1 ST - 26,50 €
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Pop-up drum fill gauge, prevent overfilling
    Pop-up drum fill gauge, prevent overfilling
    NEWPDRM317
    279,4 mm
    Log in for availability
    1 * 1 ST - 30,30 €