Order Entry
Austria
ContactUsLinkComponent
(2S,5SR)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropyl-5-methylpyrazine for synthesis, Sigma-Aldrich®
(2S,5SR)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropyl-5-methylpyrazine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
(2S,5SR)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropyl-5-methylpyrazine for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    198.26 g/mol
  • Boiling Point:
    103 - 104 °C (21 hPa)
  • Density:
    1.00 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together