4'-Nitrobenzo-18-crown-6 (4'-nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:53408-96-1
:
Formula:C₁₆H₂₃NO₈
MDL Number:MFCD00143207
:357.35 g/mol
Melting Pt:84…86 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together