2-Chloroethanesulphonic acid sodium salt hydrate for synthesis
Catalog # 8.14501.0050
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Assay (acidimetric, calc. on anhydrous substance):≥ 98.0 %
- Water (K. F.):9.0 - 11.0 %
- Identity (IR):passes test
Specification Test Results
About this item
:
:184.57 g/mol
:M-Clarity™ quality level = MQ200
EINECS:241-935-6
CAS Number:1089719-72-1
:15484-44-3
Formula:C₂H₄ClNaO₃S.H₂O
MDL Number:MFCD00007531