Order Entry
Austria
ContactUsLinkComponent
4-(2,4-Dichlorophenoxy)butyric acid methyl ester (2,4-DB methyl ester)
4-(2,4-Dichlorophenoxy)butyric acid methyl ester (2,4-DB methyl ester)
Catalog # EHERC11982000
Supplier:  EHRENSTORFER
CAS Number:  
4-(2,4-Dichlorophenoxy)butyric acid methyl ester (2,4-DB methyl ester)
Catalog # EHERC11982000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
CAS Number:18625-12-2
Formula:C₁₁H₁₂Cl₂O₃