Order Entry
Austria
ContactUsLinkComponent
 
Organic reference standard, Caffeine 100 µg/ml in aqueous solution
  CMPL398A.K1.W.L05
 :  
Organic reference standard, Caffeine 100 µg/ml in aqueous solution
  CMPL398A.K1.W.L05
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • CAS No.:
    58-08-2
  • Description:
    Caffeine 100 µg/ml in aqueous solution
  • Formula:
    C₈H₁₀N₄O₂
  • Description:
    Caffeine