Tristearin (glycerol tristearate), Sigma-Aldrich®
: Merck
:555-43-1
:
:C₅₇H₁₁₀O₆
:MFCD00036230
:891.4797 g/mol
:M-Clarity™ quality level = MQ200
:209-097-6
Frequently Bought Together
Transfer pipettes
: RATIOLAB
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Standard Line Transfer Pipettes
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection