Tristearin (glycerol tristearate) ~65%, off-white powder, Technical Grade
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:555-43-1
:
Formula:C₅₇H₁₁₀O₆
MDL Number:MFCD00036230
:891.4797 g/mol
Melting Pt:53…56 °C (lit.)
Storage Temperature:Refrigerator
EINECS:209-097-6
Frequently Bought Together