(3-Methoxyphenyl)acetic acid
Supplier: Thermo Scientific
Certificates
About this item
Synonyms: 3-Methoxyphenylacetic acid
CAS Number:1798-09-0
:
Formula:(C₆H₄(OCH₃))CH₂COOH
MDL Number:MFCD00004334
:166.18 g/mol
Boiling Pt:305,95 °C (1013 hPa)
Melting Pt:71…73 °C
Storage Temperature:Ambient
EINECS:217-282-8
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together