Order Entry
Austria
ContactUsLinkComponent
Erlenmeyer flasks, narrow or wide neck, with beaded rim
Erlenmeyer flasks, narrow or wide neck, with beaded rim
Catalog # HECH41940060
undefined
Erlenmeyer flasks, narrow or wide neck, with beaded rim
Catalog # HECH41940060

Specifications

  • Ext.Ø:
    166 mm
  • Capacity:
    2000 ml
  • Height:
    280 mm
  • Seq:
    07
  • Neck Ø ext.:
    50 mm
  • Pk:
    1

Specifications

About this item

DURAN® borosilicate glass 3.3, clear.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Volume graduations and labelling area.