ContactUsLinkComponent
Condensers, Allihn, for Soxleth extractors
Condensers, Allihn, for Soxleth extractors
Catalog # HECH41153045
undefined
Condensers, Allihn, for Soxleth extractors
Catalog # HECH41153045
Specifications
  • Cone size:
    45/40 NS
  • For:
    Soxleth extractor 100 - 250 ml
  • Pk:
    1
Specifications
About this item
DURAN® borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With NS cone. Water connection with glass nozzles.