Orders LinkContactUsLinkComponent
Laboratory bottles, narrow neck, with thread
Laboratory bottles, narrow neck, with thread
Supplier:   witeg Labortechnik
Laboratory bottles, narrow neck, with thread
Supplier:  witeg Labortechnik
Certificates
Laboratory bottles, narrow neck, with thread
Laboratory bottles, narrow neck, with thread
2 Options Available Below
Product Details & Documents
These laboratory bottles are made of borosilicate glass with safety coating witofoam.
  • Excellent chemical resistance, good heat resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Compressive strength is not increased by the plastic coating
Round, with graduation and DIN thread. The coating provides scratch, leak and splinter protection and is ideally suited to both the transport and storage of hazardous media or valuable samples.
Product Family Options
Product Information
  • Capacity
    Thread
    Availability
    Price
  • 215-1562
    Bottle with plastic coating
    Bottle with plastic coating
    20000 ml
    GL 45
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 691,00 €
    691,00 €
    1 * 1 ST
       691,00 €
  • Bottle
    5000 ml
    GL 45
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 175,00 €
    175,00 €
    1 * 1 ST
       175,00 €