ContactUsLinkComponent
Beakers, tall form
Beakers, tall form
Catalog # WITG8.601.100
Supplier:  witeg Labortechnik
Supplier #: 8.601.100
undefined
Beakers, tall form
Catalog # WITG8.601.100
Supplier:  witeg Labortechnik
Supplier #: 8.601.100
Log in for availability
Select an Option
45 mm
Select an Option
100 ml
Select an Option
80 mm
Select an Option
1
About this item
Quartz glass, with spout.
  • High thermal shock resistance with low thermal expansion
  • High transmission from the UV to the IR spectrum range
  • High chemical resistance and purity
  • High electrical insulation properties
  • High radiation resistance
  • High temperature resistance and softening temperature
The combination of the special thermal, electrical, chemical, mechanical and optical properties in a single material makes quartz glass indispensible for high-technology applications.
Frequently Bought Together