ContactUsLinkComponent
Stopcocks for desiccators
Stopcocks for desiccators
Catalog # WITG5.662.004
Supplier:  witeg Labortechnik
undefined
Stopcocks for desiccators
Catalog # WITG5.662.004
Supplier:  witeg Labortechnik
Specifications
  • Description:
    Stopcock with PTFE spindle, for desiccator lid outlet
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Stopcock, with FCH-V 24/29, flat-free high vacuum joint, made of borosilicate glass.