Orders LinkContactUsLinkComponent
Rubber Stoppers
Rubber Stoppers
Certificates
Rubber Stoppers
16 Options Available Below
Product Details & Documents
DIN 12871, made of natural rubber, hardness 40 Shore A.
Product Family Options
Without borehole
Product Information
  • No. of Holes
    Hole Ø
    Height
    Ø bottom
    Ø top
    Availability
    Price
  • 19,0 mm
    9,0 mm
    12,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 0,70 €
    0,70 €
    1 * 1 ST
       0,70 €
  • 20,0 mm
    4,0 mm
    8,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 0,50 €
    0,50 €
    1 * 1 ST
       0,50 €
  • 20,0 mm
    10,5 mm
    14,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 0,50 €
    0,50 €
    1 * 1 ST
       0,50 €
  • 20,0 mm
    14,0 mm
    18,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 0,60 €
    0,60 €
    1 * 1 ST
       0,60 €
  • 25,0 mm
    12,0 mm
    17,0 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 5,60 €
    5,60 €
    1 * 10 ST
       5,60 €
  • 25,0 mm
    17,0 mm
    22,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 0,70 €
    0,70 €
    1 * 1 ST
       0,70 €
  • 30,0 mm
    26,0 mm
    32,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1,50 €
    1,50 €
    1 * 1 ST
       1,50 €
  • 30,0 mm
    29,0 mm
    35,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1,90 €
    1,90 €
    1 * 1 ST
       1,90 €
  • 35,0 mm
    31,0 mm
    38,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,60 €
    2,60 €
    1 * 1 ST
       2,60 €
  • 35,0 mm
    34,0 mm
    41,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,80 €
    6,80 €
    1 * 1 ST
       6,80 €
  • 40,0 mm
    36,0 mm
    44,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,60 €
    3,60 €
    1 * 1 ST
       3,60 €
  • 40,0 mm
    41,0 mm
    49,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,60 €
    4,60 €
    1 * 1 ST
       4,60 €
  • 45,0 mm
    50,5 mm
    59,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,80 €
    7,80 €
    1 * 1 ST
       7,80 €
  • 45,0 mm
    56,0 mm
    65,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,30 €
    8,30 €
    1 * 1 ST
       8,30 €
  • 55,0 mm
    64,5 mm
    75,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,10 €
    10,10 €
    1 * 1 ST
       10,10 €
With borehole
Product Information
  • No. of Holes
    Hole Ø
    Height
    Ø bottom
    Ø top
    Availability
    Price
  • 2
    6,0 mm
    40,0 mm
    41,0 mm
    49,0 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 48,70 €
    48,70 €
    1 * 10 ST
       48,70 €