Order Entry
Austria
ContactUsLinkComponent
 
Desiccator cabinet, anti-static, SICCO Star
  BOHLV1910-07
 :  Bohlender
undefined
Desiccator cabinet, anti-static, SICCO Star
  BOHLV1910-07
 :  Bohlender

 

  • W×D×H ext.:
    310×375×525 mm
  • Description:
    Star-dessicator, anti-static, (42 l)
  • Environmentally Preferable:
  • Pk:
    1
  • Shelves:
    4
  • W×D×H int.:
    260×330×480 mm
  • With:
    1 chamber with 26 shelf positions, with tray hygrometer and desiccant

 

 

PC desiccator cabinet which has reinforced aluminium frame with antistatic acrylic panels and easily readable electronic hygrometer. Mainly used for temporary storage of components for electronic products.

  • Variable height telescopic shelves made of aluminium or stainless steel
  • Non-slip rubber feet or four casters (two of the casters with brakes)
  • Door with magnetic catch and circular rubber seal
  • Connection for grounding cable and tray for desiccant

These desiccators can protect electronic components against humidity and airborne particulate. Electrostatic charges are discharged by a grounding cable and a charge neutral atmosphere is produced inside the desiccator.